This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC93429 SIRNA Assays ›
Gene Symbol:
LOC93429
Gene Name:
uncharacterized LOC93429
Gene Aliases:
-
Chromosome Location:
Chr.19: 46210242 - 46214838 on Build GRCh38
UniGene ID:
Hs.569940
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n263172

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC93429
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027003.1 6 1185
GenBank mRNA AL137376.1 - 6 1184

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0