This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNHG12 SIRNA Assays ›
Gene Symbol:
SNHG12
Gene Name:
small nucleolar RNA host gene 12
Gene Aliases:
ASLNC04080, C1orf79, LINC00100, NCRNA00100
Chromosome Location:
Chr.1: 28578538 - 28581854 on Build GRCh38
UniGene ID:
Hs.632377
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n262873

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SNHG12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024127.1 1 113
GenBank mRNA AK092096.1 - 1 134
BC045792.1 - 1 131
BC064589.1 - 1 101
BC068048.1 - 1 113
KJ782215.1 - 2 1242
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CN260333.1 - 1 118 Chr. 1 - 28578597 - 28581859 [-] on Build GRCh38
GenBankmRNA BC111553.1 - 1 1391 Chr. 1 - 28581135 - 28583132 [-] on Build GRCh38
GenBankmRNA BE794361.1 - 1 93 Chr. 1 - 28578538 - 28581834 [-] on Build GRCh38
GenBankmRNA CA495298.1 - 1 118 Chr. 1 - 28579641 - 28581859 [-] on Build GRCh38
GenBankmRNA CB128668.1 - 1 132 Chr. 1 - 28579842 - 28581873 [-] on Build GRCh38
GenBankmRNA BM785231.1 - 1 131 Chr. 1 - 28579851 - 28581872 [-] on Build GRCh38
GenBankmRNA CA487963.1 - 95 Chr. - - [-] on Build
GenBankmRNA DR002536.1 - 1 124 Chr. 1 - 28579688 - 28581865 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0