This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNHG12 SIRNA Assays ›
Gene Symbol:
SNHG12
Gene Name:
small nucleolar RNA host gene 12
Gene Aliases:
ASLNC04080, C1orf79, LINC00100, NCRNA00100
Chromosome Location:
Chr.1: 28578538 - 28581854 on Build GRCh38
UniGene ID:
Hs.632377
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n262868

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SNHG12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024127.1 6 657
GenBank mRNA AF277181.1 - 6 445
AK092096.1 - 3 1826
AY277594.1 - 4 225
BC022205.1 - 6 1286
BC034611.1 - 4 1508
BC045792.1 - 5 1016
BC064589.1 - 4 1547
BC068048.1 - 6 657
BC107730.1 - 3 1049
KJ782215.1 - 7 1786
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CN260333.1 - 6 663 Chr. 1 - 28578597 - 28581859 [-] on Build GRCh38
GenBankmRNA BU075449.1 - 1 485 Chr. 1 - 28578607 - 28579104 [-] on Build GRCh38
GenBankmRNA BE794361.1 - 6 637 Chr. 1 - 28578538 - 28581834 [-] on Build GRCh38
GenBankmRNA EB387922.1 - 4 279 Chr. 1 - 28578536 - 28580649 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0