siRNA Details
Gene Symbol: URB1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026845.1 | 1 | 460 | |||
GenBank mRNA | AK026376.1 | - | 1 | 438 | ||
BC007928.1 | - | 1 | 466 |

Gene Symbol: URB1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026845.1 | 1 | 460 | |||
GenBank mRNA | AK026376.1 | - | 1 | 438 | ||
BC007928.1 | - | 1 | 466 |