This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LINC01089 SIRNA Assays ›
Gene Symbol:
LINC01089
Gene Name:
long intergenic non-protein coding RNA 1089
Gene Aliases:
-
Chromosome Location:
Chr.12: 121795267 - 121803484 on Build GRCh38
UniGene ID:
Hs.524804
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n262577

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINC01089
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002809.2 2 660
GenBank mRNA AK311480.1 - 1 216
BC038786.1 - 2 660
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA142385.1 - 1 215 Chr. 12 - 121799869 - 121802945 [-] on Build GRCh38
GenBankmRNA BF125696.1 - 2 78 Chr. 12 - 121795926 - 121802901 [-] on Build GRCh38
GenBankmRNA BI818784.1 - 2 109 Chr. 12 - 121795804 - 121802933 [-] on Build GRCh38
GenBankmRNA BG035222.1 - 161 Chr. - - [-] on Build
GenBankmRNA BE266310.1 - 1 174 Chr. 12 - 121796116 - 121802905 [-] on Build GRCh38
GenBankmRNA HY093841.1 - 1 216 Chr. 12 - 121799882 - 121802945 [-] on Build GRCh38
GenBankmRNA BG393185.1 - 2 109 Chr. 12 - 121796145 - 121802933 [-] on Build GRCh38
GenBankmRNA DC377812.1 - 1 216 Chr. 12 - 121799622 - 121802946 [-] on Build GRCh38
GenBankmRNA BI488777.1 - 99 Chr. - - [-] on Build
GenBankmRNA BI822154.1 - 2 109 Chr. 12 - 121802074 - 121802933 [-] on Build GRCh38
GenBankmRNA BM470301.1 - 102 Chr. - - [-] on Build
GenBankmRNA CN310935.1 - 1 155 Chr. 12 - 121799757 - 121802883 [-] on Build GRCh38
GenBankmRNA BM544353.1 - 130 Chr. - - [-] on Build
GenBankmRNA CN310936.1 - 1 192 Chr. 12 - 121796221 - 121802920 [-] on Build GRCh38
GenBankmRNA CD516035.1 - 2 96 Chr. 12 - 121795815 - 121802920 [-] on Build GRCh38
GenBankmRNA DA925861.1 - 2 121 Chr. 12 - 121802279 - 121802945 [-] on Build GRCh38
GenBankmRNA CN398873.1 - 2 118 Chr. 12 - 121795590 - 121802930 [-] on Build GRCh38
GenBankmRNA BM698927.1 - 1 190 Chr. 12 - 121796211 - 121802920 [-] on Build GRCh38
GenBankmRNA DA365821.1 - 1 213 Chr. 12 - 121796210 - 121802943 [-] on Build GRCh38
GenBankmRNA DA505108.1 - 215 Chr. - - [-] on Build
GenBankmRNA DT219909.1 - 1 215 Chr. 12 - 121796095 - 121802945 [-] on Build GRCh38
GenBankmRNA BG699859.1 - 2 123 Chr. 12 - 121795399 - 121802942 [-] on Build GRCh38
GenBankmRNA BX419131.2 - 1 197 Chr. 12 - 121796669 - 121802928 [-] on Build GRCh38
GenBankmRNA BX344455.2 - 2 110 Chr. 12 - 121795769 - 121802930 [-] on Build GRCh38
GenBankmRNA DT218208.1 - 2 121 Chr. 12 - 121796041 - 121802945 [-] on Build GRCh38
GenBankmRNA DB137461.1 - 2 143 Chr. 12 - 121799553 - 121802945 [-] on Build GRCh38
GenBankmRNA BE797083.1 - 2 82 Chr. 12 - 121795879 - 121802906 [-] on Build GRCh38
GenBankmRNA BX473002.1 - 1 159 Chr. 12 - 121799871 - 121802889 [-] on Build GRCh38

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AK153689.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0