This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF295-AS1 SIRNA Assays ›
Gene Symbol:
ZNF295-AS1
Gene Name:
ZNF295 antisense RNA 1
Gene Aliases:
C21orf121, NCRNA00318, PRED87
Chromosome Location:
Chr.21: 42009194 - 42024920 on Build GRCh38
UniGene ID:
Hs.143833
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n262094

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF295-AS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027273.2 2 519
NR_119384.1 2 748
NR_119385.1 2 565
GenBank mRNA AK055167.1 - 1 2579
BC029588.1 - 2 519
BI464310.1 - 2 553
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA GR939428.1 - 236 Chr. - - [-] on Build
GenBankmRNA GR939420.1 - 1 237 Chr. 21 - 42024428 - 42025180 [+] on Build GRCh38
GenBankmRNA GR939408.1 - 1 237 Chr. 21 - 42024428 - 42025154 [+] on Build GRCh38
GenBankmRNA CR744332.1 - 2 467 Chr. 21 - 42022105 - 42024712 [+] on Build GRCh38
GenBankmRNA GR939400.1 - 1 237 Chr. 21 - 42024428 - 42025028 [+] on Build GRCh38
GenBankmRNA GR937329.1 - 234 Chr. - - [-] on Build
GenBankmRNA GR939424.1 - 1 236 Chr. 21 - 42024434 - 42025161 [+] on Build GRCh38
GenBankmRNA GR939412.1 - 1 237 Chr. 21 - 42024428 - 42025192 [+] on Build GRCh38
GenBankmRNA GR939404.1 - 1 237 Chr. 21 - 42024428 - 42025180 [+] on Build GRCh38
GenBankmRNA GR939416.1 - 1 237 Chr. 21 - 42024428 - 42025206 [+] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0