This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LINC00202-1 SIRNA Assays ›
Gene Symbol:
LINC00202-1 ANKRD26
Gene Name:
long intergenic non-protein coding RNA 202-1
ankyrin repeat domain 26
Gene Aliases:
C10orf51, LINC00202, NCRNA00202, bB27G4.1
THC2, bA145E8.1
Chromosome Location:
Chr.10: 26931206 - 26942001 on Build GRCh38
Chromosome Location:
Chr.10: 26938195 - 27100498 on Build GRCh38
UniGene ID:
Hs.529965
Hs.361041
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID n262014

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINC00202-1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026795.1 1 1371
GenBank mRNA AK128027.1 - 1 2061
Gene Symbol: ANKRD26
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017015929.1 XP_016871418.1 41 22559

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header