siRNA Details
Gene Symbol: INE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002725.2 | 1 | 654 | |||
GenBank mRNA | Y10697.1 | - | 1 | 654 |

Gene Symbol: INE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002725.2 | 1 | 654 | |||
GenBank mRNA | Y10697.1 | - | 1 | 654 |