This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PIGP SIRNA Assays ›
Gene Symbol:
PIGP
Gene Name:
phosphatidylinositol glycan anchor biosynthesis class P
Gene Aliases:
DCRC, DCRC-S, DSCR5, DSRC, PIG-P
Chromosome Location:
Chr.21: 37065364 - 37073158 on Build GRCh38
UniGene ID:
Hs.656565
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n261132

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PIGP
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016430.3 NP_057514.2 3 282
XM_017028365.1 XP_016883854.1 3 224
GenBank mRNA AB037164.1 - 2 133
AF216305.1 - 3 198
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BG537429.1 - 3 230 Chr. 21 - 37065542 - 37073104 [-] on Build GRCh38
GenBankmRNA BF577360.1 - 210 Chr. - - [-] on Build
GenBankmRNA BG779865.1 - 3 159 Chr. 21 - 37065428 - 37073034 [-] on Build GRCh38
GenBankmRNA DB073689.1 - 1 22 Chr. 21 - 37071763 - 37072313 [-] on Build GRCh38
GenBankmRNA BF207636.1 - 3 208 Chr. 21 - 37065601 - 37073084 [-] on Build GRCh38
GenBankmRNA BM831879.1 - 2 46 Chr. 21 - 37065683 - 37072461 [-] on Build GRCh38
GenBankmRNA AV702373.1 - 3 149 Chr. 21 - 37065604 - 37073025 [-] on Build GRCh38
GenBankmRNA AV745310.1 - 3 160 Chr. 21 - 37065603 - 37073037 [-] on Build GRCh38
GenBankmRNA CR745554.1 - 3 204 Chr. 21 - 37065470 - 37073080 [-] on Build GRCh38
GenBankmRNA R11132.1 - 2 132 Chr. 21 - 37069562 - 37072539 [-] on Build GRCh38
GenBankmRNA BQ900325.1 - 3 142 Chr. 21 - 37065439 - 37073018 [-] on Build GRCh38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

preassembly of GPI anchor in ER membrane
phosphatidylinositol N-acetylglucosaminyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0