This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM222A-AS1 SIRNA Assays ›
Gene Symbol:
FAM222A-AS1
Gene Name:
FAM222A antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.12: 109734209 - 109773487 on Build GRCh38
UniGene ID:
Hs.675961
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n260992

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM222A-AS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026661.2 3 285
NR_026662.2 2 224
GenBank mRNA BC007821.1 - 2 225
BQ684893.1 - 2 246
DA082338.1 - 3 282
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BQ081961.1 - 2 181 Chr. 12 - 109734425 - 109773444 [-] on Build GRCh38
GenBankmRNA BF685112.1 - 217 Chr. - - [-] on Build
GenBankmRNA DA559753.1 - 1 169 Chr. 12 - 109734412 - 109735957 [-] on Build GRCh38
GenBankmRNA BI222945.1 - 3 228 Chr. 12 - 109734331 - 109773429 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0