This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ADAM3A SIRNA Assays ›
Gene Symbol:
ADAM3A
Gene Name:
ADAM metallopeptidase domain 3A (pseudogene)
Gene Aliases:
ADAM3, CYRN1, tMDCI
Chromosome Location:
Chr.8: 39451045 - 39522989 on Build GRCh38
UniGene ID:
Hs.98848
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n260420

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ADAM3A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001569.3 10 1108
NR_024106.2 10 1108
NR_024107.2 10 1108
NR_073423.1 10 1108
GenBank mRNA AK302269.1 - 7 655
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI829329.1 - 6 536 Chr. 8 - 39490262 - 39522812 [-] on Build GRCh38
GenBankmRNA CD243738.1 - 758 Chr. - - [-] on Build
GenBankmRNA DB039355.1 - 6 516 Chr. 8 - 39491979 - 39512605 [-] on Build GRCh38
GenBankmRNA DC396818.1 - 4 433 Chr. 8 - 39491911 - 39505874 [-] on Build GRCh38
GenBankmRNA BM908226.1 - 8 710 Chr. 8 - 39475427 - 39522839 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0