This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC730101 SIRNA Assays ›
Gene Symbol:
LOC730101
Gene Name:
uncharacterized LOC730101
Gene Aliases:
-
Chromosome Location:
Chr.6: 52664401 - 52669153 on Build GRCh38
UniGene ID:
Hs.7921
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n259735

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC730101
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024403.1 2 1917
NR_024405.1 3 1687
GenBank mRNA AK091117.1 - 2 1901
AK095359.1 - 2 968
AK095807.1 - 3 757

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0