This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 2010016I18RIK SIRNA Assays ›
Gene Symbol:
2010016I18Rik
Gene Name:
RIKEN cDNA 2010016I18 gene
Gene Aliases:
-
Chromosome Location:
Chr.3: 106481986 - 106485913 on Build GRCm38
UniGene ID:
Mm.14816
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n256603

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 2010016I18Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033207.1 2 203
GenBank mRNA AK079963.1 - 1 498
AK080971.1 - 2 204
AK140363.1 - 2 204
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB645704.1 - 2 204 Chr. 3 - 106482910 - 106485913 [-] on Build GRCm38
GenBankmRNA BB645667.1 - 2 204 Chr. 3 - 106482898 - 106485913 [-] on Build GRCm38
GenBankmRNA BB215580.2 - 1 242 Chr. 3 - 106482003 - 106483578 [-] on Build GRCm38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0