This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 1500015A07RIK SIRNA Assays ›
Gene Symbol:
1500015A07Rik
Gene Name:
RIKEN cDNA 1500015A07 gene
Gene Aliases:
-
Chromosome Location:
Chr.18: 61726390 - 61728254 on Build GRCm38
UniGene ID:
Mm.441992
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n256157

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1500015A07Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_029432.1 1 399
GenBank mRNA AK005240.1 - 1 399
AK133612.1 - 1 383
BC038318.1 - 356
BC047208.1 - 1 334
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BM936360.1 - 1 333 Chr. 18 - 61726456 - 61727111 [+] on Build GRCm38
GenBankmRNA CJ162399.1 - 1 373 Chr. 18 - 61726416 - 61726870 [+] on Build GRCm38
GenBankmRNA BY705419.1 - 1 399 Chr. 18 - 61726390 - 61727303 [+] on Build GRCm38
GenBankmRNA BY241015.1 - 1 366 Chr. 18 - 61726423 - 61726898 [+] on Build GRCm38
GenBankmRNA BB617829.1 - 1 383 Chr. 18 - 61726407 - 61727018 [+] on Build GRCm38
GenBankmRNA BF682564.1 - 361 Chr. - - [-] on Build
GenBankmRNA BY105078.1 - 1 383 Chr. 18 - 61726407 - 61726806 [+] on Build GRCm38
GenBankmRNA BB845143.1 - 1 382 Chr. 18 - 61726407 - 61726830 [+] on Build GRCm38
GenBankmRNA BY772473.1 - 1 366 Chr. 18 - 61726423 - 61726803 [+] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0