This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AI414108 SIRNA Assays ›
Gene Symbol:
AI414108
Gene Name:
expressed sequence AI414108
Gene Aliases:
AI854107
Chromosome Location:
Chr.9: 27352685 - 27357546 on Build GRCm38
UniGene ID:
Mm.254365
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n255955

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: AI414108
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027907.1 1 1008
GenBank mRNA AK138227.1 - 1 1009

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0