This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AI450353 SIRNA Assays ›
Gene Symbol:
AI450353
Gene Name:
expressed sequence AI450353
Gene Aliases:
-
Chromosome Location:
Chr.11: 83293430 - 83294929 on Build GRCm38
UniGene ID:
Mm.102205
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n255940

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: AI450353
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028364.1 1 795
GenBank mRNA AK134743.1 - 1 795
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY595740.1 - 1 65 Chr. 11 - 83294160 - 83294627 [+] on Build GRCm38
GenBankmRNA BF139955.1 - 1 48 Chr. 11 - 83294177 - 83294624 [+] on Build GRCm38
GenBankmRNA BX517310.1 - 1 306 Chr. 11 - 83293919 - 83294426 [+] on Build GRCm38
GenBankmRNA AA138581.1 - 1 275 Chr. 11 - 83293950 - 83294468 [+] on Build GRCm38
GenBankmRNA BY590120.1 - 1 56 Chr. 11 - 83294169 - 83294626 [+] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0