This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNHG12 SIRNA Assays ›
Gene Symbol:
Snhg12
Gene Name:
small nucleolar RNA host gene 12
Gene Aliases:
2310005L22Rik
Chromosome Location:
Chr.4: 132308678 - 132311024 on Build GRCm38
UniGene ID:
Mm.258912
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n255538

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Snhg12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_029468.1 5 469
GenBank mRNA AK009175.1 - 4 802
CB201672.1 - 5 454
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BU551930.1 - 5 374 Chr. 4 - 132308773 - 132310927 [+] on Build GRCm38
GenBankmRNA BE689861.1 - 5 429 Chr. 4 - 132308718 - 132310911 [+] on Build GRCm38
GenBankmRNA AA060788.1 - 5 334 Chr. 4 - 132308813 - 132311023 [+] on Build GRCm38
GenBankmRNA AV573991.1 - 5 398 Chr. 4 - 132308748 - 132310917 [+] on Build GRCm38
GenBankmRNA EL965250.1 - 5 435 Chr. 4 - 132308726 - 132310927 [+] on Build GRCm38
GenBankmRNA BE200523.1 - 5 446 Chr. 4 - 132308702 - 132310900 [+] on Build GRCm38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header