This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other R74862 SIRNA Assays ›
Gene Symbol:
R74862
Gene Name:
expressed sequence R74862
Gene Aliases:
-
Chromosome Location:
Chr.7: 143021784 - 143053686 on Build GRCm38
UniGene ID:
Mm.5250
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n255392

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: R74862
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015529.2 1 634
NR_110354.1 1 634
NR_110355.1 1 634
GenBank mRNA AK034864.1 - 1 626
AK035397.1 - 1 317
AK083267.1 - 1 366
BC005669.1 - 1 51
BC080794.1 - 1 634

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0