This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other B430010I23RIK SIRNA Assays ›
Gene Symbol:
B430010I23Rik
Gene Name:
RIKEN cDNA B430010I23 gene
Gene Aliases:
-
Chromosome Location:
Chr.8: 41017468 - 41024216 on Build GRCm38
UniGene ID:
Mm.440930
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n254933

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: B430010I23Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015457.1 3 1430
GenBank mRNA AK033372.1 - 3 1430
AK046663.1 - 1 1834
AK140364.1 - 2 1014

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm