siRNA Details
Gene Symbol: 1110020A21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027929.1 | 1 | 139 | |||
NR_027930.1 | 1 | 426 | ||||
GenBank mRNA | AK003841.1 | - | 1 | 142 | ||
BI737205.1 | - | 1 | 220 |

Gene Symbol: 1110020A21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027929.1 | 1 | 139 | |||
NR_027930.1 | 1 | 426 | ||||
GenBank mRNA | AK003841.1 | - | 1 | 142 | ||
BI737205.1 | - | 1 | 220 |