This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 1110020A21RIK SIRNA Assays ›
Gene Symbol:
1110020A21Rik
Gene Name:
RIKEN cDNA 1110020A21 gene
Gene Aliases:
2600015P10Rik
Chromosome Location:
Chr.17: 84917182 - 84957710 on Build GRCm38
UniGene ID:
Mm.249695
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n254687

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1110020A21Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027929.1 1 50
NR_027930.1 1 337
GenBank mRNA AK003841.1 - 1 53
BI737205.1 - 1 131
CR517132.1 - 1 346
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AA764109.1 - 1 122 Chr. 17 - 84955100 - 84957495 [-] on Build GRCm38
GenBankmRNA AI428679.1 - 1 101 Chr. 17 - 84955216 - 84957474 [-] on Build GRCm38
GenBankmRNA BX519540.1 - 1 122 Chr. 17 - 84955089 - 84957495 [-] on Build GRCm38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0