siRNA Details
Gene Symbol: Gm4566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028023.1 | 3 | 382 | |||
GenBank mRNA | CO802078.1 | - | 3 | 384 | ||
DV658461.1 | - | 2 | 504 |

Gene Symbol: Gm4566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028023.1 | 3 | 382 | |||
GenBank mRNA | CO802078.1 | - | 3 | 384 | ||
DV658461.1 | - | 2 | 504 |