This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZBTB11OS1 SIRNA Assays ›
Gene Symbol:
Zbtb11os1
Gene Name:
zinc finger and BTB domain containing 11, opposite strand 1
Gene Aliases:
2310061J03Rik
Chromosome Location:
Chr.16: 55973268 - 55974617 on Build GRCm38
UniGene ID:
Mm.472722
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n254348

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zbtb11os1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027965.1 1 977
GenBank mRNA AK006543.1 - 1 775
AK010004.1 - 1 952
BC017620.1 - 1 911 4924337
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AI390960.1 - 1 455 Chr. 16 - 55973568 - 55974095 [-] on Build GRCm38
GenBankmRNA AA013519.1 - 2 245 Chr. 16 - 55973558 - 55974584 [-] on Build GRCm38
GenBankmRNA BY578901.1 - 1 31 Chr. 16 - 55973270 - 55973668 [-] on Build GRCm38
GenBankmRNA NM_133677.1 - 911 Chr. - - [-] on Build
GenBankmRNA BU939617.1 - 70 Chr. - - [-] on Build

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0