This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM120AOS SIRNA Assays ›
Gene Symbol:
Fam120aos
Gene Name:
family with sequence similarity 120A, opposite strand
Gene Aliases:
C030044B11Rik
Chromosome Location:
Chr.13: 48968112 - 48969905 on Build GRCm38
UniGene ID:
Mm.426304
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n253241

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam120aos
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015601.1 2 549
GenBank mRNA AK014080.1 - 2 279
AK021143.1 - 2 241
BC024672.1 - 374
BC045201.1 - 2 481
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI143388.1 - 2 277 Chr. 13 - 48968385 - 48969479 [+] on Build GRCm38
GenBankmRNA BY728294.1 - 2 240 Chr. 13 - 48968422 - 48969418 [+] on Build GRCm38
GenBankmRNA AA024302.1 - 2 240 Chr. 13 - 48968418 - 48969113 [+] on Build GRCm38
GenBankmRNA AW213542.1 - 2 218 Chr. 13 - 48968443 - 48969229 [+] on Build GRCm38
GenBankmRNA BY231388.1 - 2 310 Chr. 13 - 48968351 - 48969054 [+] on Build GRCm38
GenBankmRNA BF167442.1 - 2 370 Chr. 13 - 48968293 - 48970987 [+] on Build GRCm38
GenBankmRNA BU850808.1 - 273 Chr. - - [-] on Build
GenBankmRNA BY713078.1 - 2 276 Chr. 13 - 48968385 - 48969165 [+] on Build GRCm38
GenBankmRNA BI962234.1 - 2 269 Chr. 13 - 48968392 - 48969108 [+] on Build GRCm38
GenBankmRNA BM937943.1 - 2 220 Chr. 13 - 48968441 - 48969271 [+] on Build GRCm38
GenBankmRNA BG976926.1 - 2 272 Chr. 13 - 48968389 - 48969436 [+] on Build GRCm38
GenBankmRNA BQ936809.1 - 2 157 Chr. 13 - 48968504 - 48969402 [+] on Build GRCm38
GenBankmRNA BI738904.1 - 2 480 Chr. 13 - 48968181 - 48969249 [+] on Build GRCm38
GenBankmRNA AA764629.1 - 2 281 Chr. 13 - 48968389 - 48969230 [+] on Build GRCm38
GenBankmRNA CN842812.1 - 266 Chr. - - [-] on Build
GenBankmRNA BX522417.1 - 2 337 Chr. 13 - 48968324 - 48969207 [+] on Build GRCm38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0