This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM120AOS SIRNA Assays ›
Gene Symbol:
Fam120aos
Gene Name:
family with sequence similarity 120A, opposite strand
Gene Aliases:
C030044B11Rik
Chromosome Location:
Chr.13: 48968112 - 48969905 on Build GRCm38
UniGene ID:
Mm.426304
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n253238

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam120aos
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015601.1 2 933
GenBank mRNA AK021143.1 - 2 625
BC045201.1 - 2 865
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BQ936809.1 - 2 541 Chr. 13 - 48968504 - 48969402 [+] on Build GRCm38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0