This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other D2WSU81E SIRNA Assays ›
Gene Symbol:
D2Wsu81e
Gene Name:
DNA segment, Chr 2, Wayne State University 81, expressed
Gene Aliases:
9930028P05, AL033358
Chromosome Location:
Chr.2: 30173447 - 30178495 on Build GRCm38
UniGene ID:
Mm.4449
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n252552

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: D2Wsu81e
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK044983.1 - 1 181
BY289626.1 - 1 199
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BU514809.1 - 1 40 Chr. 2 - 30176375 - 30178290 [-] on Build GRCm38
GenBankmRNA CJ146774.1 - 1 191 Chr. 2 - 30177978 - 30178440 [-] on Build GRCm38
GenBankmRNA CJ146684.1 - 1 191 Chr. 2 - 30177971 - 30178440 [-] on Build GRCm38
GenBankmRNA BB643208.1 - 1 181 Chr. 2 - 30177974 - 30178431 [-] on Build GRCm38
GenBankmRNA BU510712.1 - 1 193 Chr. 2 - 30177518 - 30178415 [-] on Build GRCm38
GenBankmRNA BG175931.1 - 1 47 Chr. 2 - 30177332 - 30178297 [-] on Build GRCm38
GenBankmRNA BB618850.1 - 1 206 Chr. 2 - 30177663 - 30178455 [-] on Build GRCm38
GenBankmRNA CX229350.1 - 1 180 Chr. 2 - 30177648 - 30178430 [-] on Build GRCm38
GenBankmRNA CJ146801.1 - 1 191 Chr. 2 - 30177959 - 30178440 [-] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header