This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNHG6 SIRNA Assays ›
Gene Symbol:
Snhg6
Gene Name:
small nucleolar RNA host gene 6
Gene Aliases:
1110008H02Rik
Chromosome Location:
Chr.1: 9942025 - 9944118 on Build GRCm38
UniGene ID:
Mm.28311
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n251643

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Snhg6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024067.2 2 190
GenBank mRNA AK003573.1 - 2 191
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA W71553.1 - 167 Chr. - - [-] on Build
GenBankmRNA BY043991.1 - 2 190 Chr. 1 - 9942072 - 9944117 [-] on Build GRCm38
GenBankmRNA BY788907.1 - 2 189 Chr. 1 - 9942074 - 9944117 [-] on Build GRCm38
GenBankmRNA CJ082230.1 - 2 189 Chr. 1 - 9942037 - 9944117 [-] on Build GRCm38
GenBankmRNA AA015539.1 - 2 150 Chr. 1 - 9942042 - 9944079 [-] on Build GRCm38
GenBankmRNA AA023387.1 - 2 152 Chr. 1 - 9942023 - 9944083 [-] on Build GRCm38
GenBankmRNA BY604390.1 - 2 195 Chr. 1 - 9942019 - 9944118 [-] on Build GRCm38
GenBankmRNA BE853719.1 - 2 175 Chr. 1 - 9942153 - 9944103 [-] on Build GRCm38
GenBankmRNA BI109518.1 - 2 173 Chr. 1 - 9942023 - 9944099 [-] on Build GRCm38
GenBankmRNA AV468093.1 - 2 150 Chr. 1 - 9942028 - 9944077 [-] on Build GRCm38
GenBankmRNA AA097989.1 - 2 160 Chr. 1 - 9942025 - 9944095 [-] on Build GRCm38
GenBankmRNA BE134757.1 - 2 170 Chr. 1 - 9942023 - 9944098 [-] on Build GRCm38
GenBankmRNA BX523010.1 - 2 180 Chr. 1 - 9942023 - 9944107 [-] on Build GRCm38
GenBankmRNA W35693.1 - 119 Chr. - - [-] on Build
GenBankmRNA W18398.1 - 159 Chr. - - [-] on Build
GenBankmRNA AA940467.1 - 2 169 Chr. 1 - 9941959 - 9944090 [-] on Build GRCm38
GenBankmRNA BX516798.1 - 2 180 Chr. 1 - 9942023 - 9944105 [-] on Build GRCm38
GenBankmRNA AL034794.1 - 1 280 Chr. 1 - 9942230 - 9942897 [-] on Build GRCm38
GenBankmRNA AA002823.1 - 2 173 Chr. 1 - 9942023 - 9944098 [-] on Build GRCm38
GenBankmRNA AA289428.1 - 1 75 Chr. 1 - 9942025 - 9942695 [-] on Build GRCm38
GenBankmRNA AA200900.1 - 2 154 Chr. 1 - 9942040 - 9944086 [-] on Build GRCm38
GenBankmRNA AA155393.1 - 2 115 Chr. 1 - 9942025 - 9944043 [-] on Build GRCm38
GenBankmRNA AA124208.1 - 2 174 Chr. 1 - 9942058 - 9944104 [-] on Build GRCm38
GenBankmRNA BY346229.1 - 2 193 Chr. 1 - 9942118 - 9944121 [-] on Build GRCm38
GenBankmRNA BY711012.1 - 2 189 Chr. 1 - 9941960 - 9944117 [-] on Build GRCm38
GenBankmRNA CK793285.1 - 173 Chr. - - [-] on Build
GenBankmRNA AV490092.1 - 2 154 Chr. 1 - 9942038 - 9944081 [-] on Build GRCm38
GenBankmRNA BE624330.1 - 2 152 Chr. 1 - 9942023 - 9944080 [-] on Build GRCm38
GenBankmRNA CJ320246.1 - 1 57 Chr. 1 - 9942026 - 9942683 [-] on Build GRCm38
GenBankmRNA AV115890.2 - 369 Chr. - - [-] on Build
GenBankmRNA AA250387.1 - 2 167 Chr. 1 - 9942054 - 9944087 [-] on Build GRCm38
GenBankmRNA AV485517.1 - 2 177 Chr. 1 - 9942023 - 9944104 [-] on Build GRCm38
GenBankmRNA BY345355.1 - 2 193 Chr. 1 - 9942107 - 9944121 [-] on Build GRCm38
GenBankmRNA BY111725.1 - 2 189 Chr. 1 - 9942091 - 9944117 [-] on Build GRCm38
GenBankmRNA AA267148.1 - 1 28 Chr. 1 - 9942023 - 9942654 [-] on Build GRCm38
GenBankmRNA AA023086.1 - 2 179 Chr. 1 - 9942024 - 9944105 [-] on Build GRCm38
GenBankmRNA AA119805.1 - 2 165 Chr. 1 - 9942023 - 9944098 [-] on Build GRCm38
GenBankmRNA W40695.1 - 118 Chr. - - [-] on Build
GenBankmRNA W54219.1 - 115 Chr. - - [-] on Build
GenBankmRNA BY345466.1 - 2 193 Chr. 1 - 9942116 - 9944121 [-] on Build GRCm38
GenBankmRNA W11261.1 - 173 Chr. - - [-] on Build
GenBankmRNA W98878.1 - 168 Chr. - - [-] on Build
GenBankmRNA BY034531.1 - 2 190 Chr. 1 - 9942060 - 9944117 [-] on Build GRCm38
GenBankmRNA AA146116.1 - 2 110 Chr. 1 - 9942033 - 9944038 [-] on Build GRCm38
GenBankmRNA AW988981.1 - 2 170 Chr. 1 - 9942023 - 9944098 [-] on Build GRCm38
GenBankmRNA AA098159.1 - 2 167 Chr. 1 - 9942037 - 9944097 [-] on Build GRCm38
GenBankmRNA AA014503.1 - 2 131 Chr. 1 - 9942168 - 9944058 [-] on Build GRCm38
GenBankmRNA BY703829.1 - 2 189 Chr. 1 - 9942025 - 9944117 [-] on Build GRCm38
GenBankmRNA BF715116.1 - 2 177 Chr. 1 - 9942023 - 9944105 [-] on Build GRCm38
GenBankmRNA AA240411.1 - 1 28 Chr. 1 - 9942039 - 9942654 [-] on Build GRCm38
GenBankmRNA W54183.1 - 172 Chr. - - [-] on Build
GenBankmRNA AV115922.2 - 2 211 Chr. 1 - 9941973 - 9944118 [-] on Build GRCm38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0