This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GM17801 SIRNA Assays ›
Gene Symbol:
Gm17801
Gene Name:
predicted gene, 17801
Gene Aliases:
-
Chromosome Location:
Chr.17: 25274124 - 25275528 on Build GRCm38
UniGene ID:
Mm.478766
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n251627

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm17801
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027452.1 1 1383
GenBank mRNA AK143641.1 - 1 1384
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB735807.1 - 1 384 Chr. 17 - 25275123 - 25275528 [+] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0