This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 4930558C23RIK SIRNA Assays ›
Gene Symbol:
4930558C23Rik
Gene Name:
RIKEN cDNA 4930558C23 gene
Gene Aliases:
-
Chromosome Location:
Chr.3: 95386458 - 95402841 on Build GRCm38
UniGene ID:
Mm.75066
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n251529

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4930558C23Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015490.1 2 378
GenBank mRNA AK006566.1 - 2 345
AK016169.1 - 2 369
BC049745.1 - 2 378

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0