This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MIRG SIRNA Assays ›
Gene Symbol:
Mirg
Gene Name:
miRNA containing gene
Gene Aliases:
2810474H01Rik, AI852992, Gm2928, Meg9, mir-410
Chromosome Location:
Chr.12: 109734981 - 109749458 on Build GRCm38
UniGene ID:
Mm.260543
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n251315

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mirg
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028265.1
2 108
GenBank mRNA AI605771.1 -
2 108
AJ517767.1 - LIT1749
Mirg
2 108
EU434920.1 -
1855
EU616812.1 -
2692
EU616813.1 -
2605
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CB193946.1 - 2 104 Chr. 12 - 109734985 - 109743827 [+] on Build GRCm38
GenBankmRNA CB193969.1 - 2 104 Chr. 12 - 109734985 - 109748183 [+] on Build GRCm38
GenBankmRNA BG805738.1 - 2 93 Chr. 12 - 109734996 - 109742244 [+] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0