This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 4933431E20RIK SIRNA Assays ›
Gene Symbol:
4933431E20Rik
Gene Name:
RIKEN cDNA 4933431E20 gene
Gene Aliases:
-
Chromosome Location:
Chr.3: 107888850 - 107896213 on Build GRCm38
UniGene ID:
Mm.270500
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n251294

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4933431E20Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015459.1 3 2758
GenBank mRNA AK030251.1 - 4 2380
AK036227.1 - 3 2331
AK147824.1 - 4 2355
BC060136.1 - 3 2758
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA NM_177858.1 - 2331 Chr. - - [-] on Build

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header