This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other STAT4 SIRNA Assays ›
Gene Symbol:
STAT4
Gene Name:
signal transducer and activator of transcription 4
Gene Aliases:
SLEB11
Chromosome Location:
Chr.2: 191029576 - 191172684 on Build GRCh38
UniGene ID:
Hs.735572
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID VHS41329
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: STAT4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243835.1 NP_001230764.1 17, 18 1829
NM_003151.3 NP_003142.1 17, 18 1880
XM_006712719.3 XP_006712782.1 17, 18 1875
XM_011511705.2 XP_011510007.1 20, 21 2026
GenBank mRNA AB209043.1 - 7, 8 516
AK299976.1 - 14, 15 1361
AK300868.1 - 17, 18 1633
AK301172.1 - 15, 16 1304
BC031212.1 - 17, 18 1801 4830583
L78440.1 - 17, 18 1646

Back To Top

More Information


Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
JAK-STAT cascade
RNA polymerase II regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0