This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DEAF1 SIRNA Assays ›
Gene Symbol:
Deaf1
Gene Name:
DEAF1 transcription factor
Gene Aliases:
-
Chromosome Location:
Chr.1: 214283787 - 214317466 on Build Rnor_6.0
UniGene ID:
Rn.11134
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS372990
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Deaf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031801.1 NP_113989.1 4 665
XM_006230582.2 XP_006230644.1 4 1533
XM_006230583.2 XP_006230645.1 4 1533
XM_008760011.1 XP_008758233.1 4 584
XM_008760012.1 XP_008758234.1 4 584
XM_008760013.1 XP_008758235.1 4 584
XM_008760014.1 XP_008758236.1 4 1549
GenBank mRNA AF055884.1 - 4 665
U59659.1 - 4 414

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001282073, NM_001282072, NM_016874, NM_001282076, AK158586, and others.

Gene Ontology Categories:

Function(s) Process(es)

behavioral fear response
neural tube closure
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
visual learning
regulation of mammary gland epithelial cell proliferation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic skeletal system development
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0