This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GRK1 SIRNA Assays ›
Gene Symbol:
Grk1
Gene Name:
G protein-coupled receptor kinase 1
Gene Aliases:
Rhok
Chromosome Location:
Chr.16: 81153438 - 81169128 on Build Rnor_6.0
UniGene ID:
Rn.10548
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS372867
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Grk1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031096.1 NP_112358.1 7 1696
GenBank mRNA U63971.1 - 7 1696

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction
visual perception
photoreceptor cell morphogenesis
response to light stimulus
rhodopsin mediated signaling pathway
regulation of rhodopsin mediated signaling pathway
termination of G-protein coupled receptor signaling pathway
positive regulation of phosphorylation
response to drug
negative regulation of apoptotic process
protein autophosphorylation
post-embryonic retina morphogenesis in camera-type eye
G-protein coupled receptor kinase activity
ATP binding
rhodopsin kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header