This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LSS SIRNA Assays ›
Gene Symbol:
Lss
Gene Name:
lanosterol synthase (2,3-oxidosqualene-lanosterol cyclase)
Gene Aliases:
-
Chromosome Location:
Chr.20: 12844522 - 12870474 on Build Rnor_6.0
UniGene ID:
Rn.10211
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS372835
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lss
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031049.1 NP_112311.1 16, 17 1609
GenBank mRNA D45252.1 - 16, 17 1633
U31352.1 - 16, 17 1609

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_146006, AK044016, AK154520, BC029082, AK054393, and others.

Gene Ontology Categories:

Function(s) Process(es)

cholesterol biosynthetic process
metabolic process
sterol metabolic process
lanosterol synthase activity
intramolecular transferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0