This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF168 SIRNA Assays ›
Gene Symbol:
Rnf168
Gene Name:
ring finger protein 168, E3 ubiquitin protein ligase
Gene Aliases:
-
Chromosome Location:
Chr.11: 71721242 - 71743421 on Build Rnor_6.0
UniGene ID:
Rn.101441
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS372144
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rnf168
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127597.2 NP_001121069.2 6 1025
GenBank mRNA BC166869.1 - 6 1025 9030307

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair
ubiquitin-dependent protein catabolic process
cellular response to DNA damage stimulus
biological_process
response to ionizing radiation
protein ubiquitination
negative regulation of transcription elongation from RNA polymerase II promoter
histone H2A monoubiquitination
histone H2A-K13 ubiquitination
histone H2A-K15 ubiquitination
isotype switching
positive regulation of DNA repair
protein K63-linked ubiquitination
histone H2A K63-linked ubiquitination
molecular_function
chromatin binding
ubiquitin-protein transferase activity
zinc ion binding
ligase activity
nucleosome binding
histone binding
ubiquitin binding
K63-linked polyubiquitin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0