siRNA Details
Gene Symbol: Gba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127639.1 | NP_001121111.1 | 8 | 1124 | ||
GenBank mRNA | BC166732.1 | - | 8 | 1124 | 7934870 |

Gene Symbol: Gba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127639.1 | NP_001121111.1 | 8 | 1124 | ||
GenBank mRNA | BC166732.1 | - | 8 | 1124 | 7934870 |