This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CHCHD7 SIRNA Assays ›
Gene Symbol:
Chchd7
Gene Name:
coiled-coil-helix-coiled-coil-helix domain containing 7
Gene Aliases:
-
Chromosome Location:
Chr.5: 16843246 - 16847809 on Build Rnor_6.0
UniGene ID:
-
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS370205
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Chchd7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749892.3 XP_003749940.1 5 626
XM_003749893.2 XP_003749941.1 5 465
XM_008763563.1 XP_008761785.1 5 688

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001190323, NM_001190324, NM_001190322, NM_001285804, NM_181391, and others.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0