siRNA Details
Gene Symbol: Chchd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749892.3 | XP_003749940.1 | 3, 4 | 476 | ||
XM_003749893.2 | XP_003749941.1 | 3, 4 | 315 |

Gene Symbol: Chchd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749892.3 | XP_003749940.1 | 3, 4 | 476 | ||
XM_003749893.2 | XP_003749941.1 | 3, 4 | 315 |