This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF13 SIRNA Assays ›
Gene Symbol:
Rnf13
Gene Name:
ring finger protein 13
Gene Aliases:
-
Chromosome Location:
Chr.2: 147853423 - 147948277 on Build Rnor_6.0
UniGene ID:
Rn.17153
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS369176
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rnf13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109444.2 NP_001102914.1 920
GenBank mRNA BC081881.1 - 920 7108181
FQ224754.1 - 1088

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001113413, BC058182, AK158046, AF037205, AK167278, and others.

Gene Ontology Categories:

Function(s) Process(es)

protein autoubiquitination
ubiquitin-protein transferase activity
zinc ion binding
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm