This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GNG11 SIRNA Assays ›
Gene Symbol:
Gng11 LOC100912034
Gene Name:
guanine nucleotide binding protein (G protein), gamma 11
guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-11-like
Gene Aliases:
-
-
Chromosome Location:
Chr.4: 28989170 - 28994610 on Build Rnor_6.0
Chromosome Location:
Chr.4: 31387262 - 31392791 on Build Rnor_6.0
UniGene ID:
Rn.892
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID RSS368047
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gng11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022396.2 NP_071791.1 1, 2 406
GenBank mRNA AF257110.1 - 1, 2 127
BC097320.1 - 1, 2 406 7455035
Gene Symbol: LOC100912034
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749698.3 XP_003749746.1 1, 2 564

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
biological_process
molecular_function
signal transducer activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header