This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BIRC5 SIRNA Assays ›
Gene Symbol:
Birc5
Gene Name:
baculoviral IAP repeat-containing 5
Gene Aliases:
AP14
Chromosome Location:
Chr.10: 106856097 - 106864419 on Build Rnor_6.0
UniGene ID:
Rn.54471
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS367925
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Birc5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022274.1 NP_071610.1 1 35
GenBank mRNA AF276775.1 - 1 35

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
microtubule cytoskeleton organization
transcription, DNA-templated
protein phosphorylation
chromosome segregation
regulation of mitotic cell cycle
regulation of signal transduction
protein ubiquitination
protein complex localization
positive regulation of exit from mitosis
cellular response to stem cell factor stimulus
response to drug
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
response to peptide hormone
negative regulation of neuron apoptotic process
positive regulation of cell cycle
negative regulation of transcription, DNA-templated
positive regulation of mitotic cell cycle
cell division
establishment of chromosome localization
response to glucocorticoid
regulation of insulin secretion involved in cellular response to glucose stimulus
regulation of type B pancreatic cell proliferation
cellular response to ethanol
mitotic spindle assembly
inhibition of cysteine-type endopeptidase activity involved in apoptotic process
molecular_function
ubiquitin-protein transferase activity
cysteine-type endopeptidase inhibitor activity
microtubule binding
zinc ion binding
Ran GTPase binding
tubulin binding
enzyme binding
identical protein binding
protein homodimerization activity
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
protein heterodimerization activity
cofactor binding
chaperone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0