This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PTGES SIRNA Assays ›
Gene Symbol:
Ptges
Gene Name:
prostaglandin E synthase
Gene Aliases:
Pges
Chromosome Location:
Chr.3: 9727408 - 9738752 on Build Rnor_6.0
UniGene ID:
Rn.7730
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS367656
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ptges
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021583.3 NP_067594.1 1 154
GenBank mRNA AB035145.1 - 1 114
AB041998.1 - 1 143
AB048730.1 - 1 156
AF280967.1 - 1 153
BC088101.1 - 1 112
BC105858.1 - 1 140 7456259

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

prostaglandin biosynthetic process
acute inflammatory response
chronic inflammatory response
prostaglandin metabolic process
negative regulation of cell proliferation
response to organic cyclic compound
response to lipopolysaccharide
response to retinoic acid
response to cytokine
response to calcium ion
glutathione binding
prostaglandin-E synthase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0