This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NDUFAF3 SIRNA Assays ›
Gene Symbol:
Ndufaf3
Gene Name:
NADH dehydrogenase (ubiquinone) complex I, assembly factor 3
Gene Aliases:
RGD708545
Chromosome Location:
Chr.8: 117351628 - 117355416 on Build Rnor_6.0
UniGene ID:
Rn.40118
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS367538
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ndufaf3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033971.1 NP_001029143.1 4 1049
NM_020080.2 NP_064465.1 4 1067
XM_006243845.2 XP_006243907.1 5 2549
GenBank mRNA BC079306.1 - 4 801 7134644
FQ216095.1 - 4 418
FQ223486.1 - 3 495
FQ233128.1 - 3 494
U95160.1 - 4 403
U95161.1 - 4 469
U95162.1 - 3 573

Back To Top

More Information


Gene Ontology Categories:

Function(s)

mitochondrial respiratory chain complex I assembly

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0