This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FURIN SIRNA Assays ›
Gene Symbol:
Furin
Gene Name:
furin (paired basic amino acid cleaving enzyme)
Gene Aliases:
Pace, Pcsk3
Chromosome Location:
Chr.1: 142185092 - 142198166 on Build Rnor_6.0
UniGene ID:
Rn.3220
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS367447
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Furin
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019331.1 NP_062204.1 5 930
XM_008759555.1 XP_008757777.1 5 814
XM_008759556.1 XP_008757778.1 5 861
GenBank mRNA X55660.1 - 5 930

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal peptide processing
proteolysis
aging
cell proliferation
regulation of signal transduction
negative regulation of endopeptidase activity
protein processing
peptide hormone processing
viral life cycle
positive regulation of cell migration
positive regulation of transforming growth factor beta receptor signaling pathway
negative regulation of low-density lipoprotein particle receptor catabolic process
nerve growth factor production
negative regulation of nerve growth factor production
negative regulation of transforming growth factor beta1 production
secretion by cell
regulation of protein catabolic process
peptide biosynthetic process
regulation of low-density lipoprotein particle receptor biosynthetic process
regulation of endopeptidase activity
protease binding
endopeptidase activity
serine-type endopeptidase activity
serine-type endopeptidase inhibitor activity
peptidase activity
peptide binding
metal ion binding
nerve growth factor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0