siRNA Details
Gene Symbol: Clec9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109354.1 | NP_001102824.1 | 4 | 508 | ||
GenBank mRNA | GU357486.1 | - | 4 | 412 |

Gene Symbol: Clec9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109354.1 | NP_001102824.1 | 4 | 508 | ||
GenBank mRNA | GU357486.1 | - | 4 | 412 |