This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CLEC9A SIRNA Assays ›
Gene Symbol:
Clec9a
Gene Name:
C-type lectin domain family 9, member A
Gene Aliases:
RGD1562513
Chromosome Location:
Chr.4: 163173597 - 163189532 on Build Rnor_6.0
UniGene ID:
Rn.14822
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS367119
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Clec9a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109354.1 NP_001102824.1 2, 3 274
XM_006237099.2 XP_006237161.1 2, 3 1150
XM_008763325.1 XP_008761547.1 1, 2 452
XM_008763326.1 XP_008761548.1 2, 3 151
GenBank mRNA GU357486.1 - 2, 3 178

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

receptor-mediated endocytosis
positive regulation of cytokine secretion
carbohydrate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0