This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TBC1D5 SIRNA Assays ›
Gene Symbol:
Tbc1d5
Gene Name:
TBC1 domain family, member 5
Gene Aliases:
RGD1561626
Chromosome Location:
Chr.9: 1144644 - 1747732 on Build Rnor_6.0
UniGene ID:
Rn.203950
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS366283
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tbc1d5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134762.1 NP_001128234.1 22 2467
XM_008766768.1 XP_008764990.1 23 3589
XM_008766769.1 XP_008764991.1 19 2085

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of receptor internalization
positive regulation of autophagy
retrograde transport, endosome to Golgi
response to starvation
regulation of cilium assembly
AP-2 adaptor complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0