This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LRRTM4 SIRNA Assays ›
Gene Symbol:
Lrrtm4
Gene Name:
leucine rich repeat transmembrane neuronal 4
Gene Aliases:
RGD1560707
Chromosome Location:
Chr.4: 110699580 - 111497643 on Build Rnor_6.0
UniGene ID:
Rn.140910
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS365373
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lrrtm4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134746.1 NP_001128218.1 1 1007
XM_008763024.1 XP_008761246.1 2 1326
XM_008763025.1 XP_008761247.1 3 1479
XM_008763026.1 XP_008761248.1 3 1390
GenBank mRNA BC168219.1 - 1 1007 7319359

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_178731, NM_001282102, NM_001134743, AK032470, AK038656, and others.

Gene Ontology Categories:

Function(s) Process(es)

synapse organization
neurotransmitter-gated ion channel clustering
AMPA glutamate receptor clustering
regulation of presynaptic membrane organization
heparan sulfate proteoglycan binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0