This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EVX1 SIRNA Assays ›
Gene Symbol:
Evx1 NEWGENE_1565788
Gene Name:
even-skipped homeobox 1
even-skipped homeobox 1
Gene Aliases:
RGD1565788
Evx1
Chromosome Location:
Chr.4|NW_007906169: 186548 - 190925 on Build Rnor_6.0
Chromosome Location:
Chr.4: 82370986 - 82374313 on Build Rnor_6.0
UniGene ID:
Rn.145628
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID RSS365322
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Evx1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191972.1 NP_001178901.1 3 1109
Gene Symbol: NEWGENE_1565788
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762952.1 XP_008761174.1 3 1120

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007966, X54239, BC126907.

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm